missing translation for 'onlineSavingsMsg'
Learn More

Cuprous Chloride, ≥90%, (ACS Reagent Grade), MP Biomedicals

Product Code. 11442822 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11442822 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11442822 Supplier MP Biomedicals Supplier No. 0215251580

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 7758-89-6
Quantity 100 g
Assay Percent Range ≥90%
Molecular Formula ClCu
Synonym cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech
InChI Key OXBLHERUFWYNTN-UHFFFAOYSA-M
SMILES Cl[Cu]
IUPAC Name chlorocopper
Molecular Weight (g/mol) 98.996
PubChem CID 62652
ChEBI CHEBI:53472
Formula Weight 99
Grade ACS Reagent
Chemical Name or Material Cuprous Chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.