missing translation for 'onlineSavingsMsg'
Learn More

Cupric Sulfate, Anhydrous, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15597924 Shop All J.T.Baker Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
Packaging:
Poly Bottle
missing translation for 'unitSize':
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging missing translation for 'unitSize'
15597924 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 15597924 missing translation for 'mfr' Avantor J.T.Baker missing translation for 'supplierNo' 185001

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 7758-98-7
Physical Form Powder
Quantity 500 g
Molecular Formula CuO4S
Synonym copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagle
InChI Key ARUVKPQLZAKDPS-UHFFFAOYSA-L
SMILES [O-]S(=O)(=O)[O-].[Cu+2]
IUPAC Name copper;sulfate
Molecular Weight (g/mol) 159.602
PubChem CID 24462
ChEBI CHEBI:23414
Formula Weight 159.6
Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Specific Gravity 1L = 3.60kg
Chemical Name or Material Cupric Sulfate, Anhydrous
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.