missing translation for 'onlineSavingsMsg'
Learn More

Cupric Nitrate Hydrate, MP Biomedicals

Product Code. 11428801 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428801 100 g 100g
11438801 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11428801 Supplier MP Biomedicals Supplier No. 0215350180
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 10031-43-3
Molecular Formula CuH6N2O9
Molecular Weight (g/mol) 241.599
InChI Key SXTLQDJHRPXDSB-UHFFFAOYSA-N
Synonym copper ii nitrate trihydrate, copper nitrate trihydrate, cupric nitrate trihydrate, copper ii nitrate hydrate, copper ii nitrate 3-hydrate, copper 2+ trihydrate dinitronate, copper 2+ trihydrate dinitrate, copper ii nitrate trihydrate-cu puratrem
PubChem CID 9837674
IUPAC Name copper;dinitrate;trihydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.[Cu+2]

Specifications

Quantity 100 g
Formula Weight 187.6 + aq.
Chemical Name or Material Cupric Nitrate Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.