missing translation for 'onlineSavingsMsg'
Learn More

Cupri-Tartaric (I+II) solution EU Pharmacopoeia, Fisher Chemical™
BIOPHARMA

Product Code. 12980394 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12980394 1000 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12980394 Supplier Fisher Chemical Supplier No. RP 060 500
  • ONLINE EXCLUSIVE PRICE
    £352.00
    £240.16 / 500mL
    Save £111.84
    31% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

Packaging Bottle
CAS 77-92-9
Molecular Formula C6H8O7
MDL Number MFCD00011669
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
Molecular Weight (g/mol) 192.12
PubChem CID 311
Quantity 1000 mL
ChEBI CHEBI:30769
Grade EU Ph Ref: 1023100
Chemical Name or Material Cupri-citric solution
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cupri-tartaric solution. (Ready to use Mixture) European Pharmacopoeia Ref: 1023300 (RP 060)
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.