missing translation for 'onlineSavingsMsg'
Learn More

Cupferron, Certified AR for Analysis, Fisher Chemical

Product Code. 11478133 Shop All Fisher Chemical Products
Change view
Click to view available options
100g, Glass Jar
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11478133 100g, Glass Jar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11478133 Supplier Fisher Chemical Supplier No. C/7720/48
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 135-20-6
Melting Point 150°C
Color Beige
Packaging Glass Jar
Molecular Formula C6H9N3O2
MDL Number MFCD00078422
Quantity 100 g
InChI Key NJVHCUNZAMFQNA-UHFFFAOYSA-N
SMILES N.ON(N=O)C1=CC=CC=C1
IUPAC Name N-nitroso-N-phenylhydroxylamine amine
Molecular Weight (g/mol) 155.16
Formula Weight 155.16g/mol
Physical Form Solid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cupferron
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.