missing translation for 'onlineSavingsMsg'
Learn More

Key Industrial Equipment™ Cupboard for general use with four shelves, grey/blue, 1950mm x 1000mm x 450mm

Brand:  Key Industrial Equipment™ A334475

Encompass_Preferred

Product Code. 12854988

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Durable, heavy duty storage cupboards with extra strong welded door frames for greater security. Concealed hinges for a clean line appearance and a wipe-clean finish with a tough protective coating. Fitted lock supplied with two keys.
  • Blue door
  • Four shelves
  • Dimensions (H x W x D) : 1950mm x 1000mm x 450mm
  • Shelf capacity UDL : 100kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.