Learn More
OHAUS™ Cup Head
OHAUS™ Cup Head is an accessory for use with a vortex mixer.
Brand:  OHAUS™ 30400235
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15772649
  • £40.20 / Each
Estimated Shipment: 28-08-2024
to see stock.
Add to basket
Description
Description
  • Increase throughput by mixing multiple samples at one time
  • Effortlessly mix up to 52 microtubes or eight microplates
  • Choose the appropriate holder for the sample size, or select the flat insert and cut out to suit custom specifications
Specifications
Specifications
Cup Head
Black
Vortex Mixers
Rubber
Cup Head
1 Ea.