missing translation for 'onlineSavingsMsg'
Learn More

Cumyl hydroperoxide, 80%

Product Code. 11934761 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
2.5kg
250g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11934761 2.5 kg Plastic bottle 2.5kg
10728732 5 g Plastic bottle 5g
10336451 250 g Plastic bottle 250g
10219373 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11934761 Supplier Thermo Scientific Acros Supplier No. 349960025
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 80-15-9 | C9H12O2 | 152.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 80-15-9 , 98-82-8 , 617-94-7 , 98-86-2
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00002129
InChI Key YQHLDYVWEZKEOX-UHFFFAOYSA-N
Synonym cumene hydroperoxide, cumyl hydroperoxide, hydroperoxide, 1-methyl-1-phenylethyl, cumenyl hydroperoxide, alpha,alpha-dimethylbenzyl hydroperoxide, cumolhydroperoxid, 7-cumyl hydroperoxide, cumolhydroperoxide, cument hydroperoxide, hydroperoxyde de cumene
PubChem CID 6629
ChEBI CHEBI:78673
SMILES CC(C)(OO)C1=CC=CC=C1

Spécification

Melting Point -30.0°C
Density 1.0600g/mL
Flash Point 83°C
Assay Percent Range 80 to 85% (Iodometry)
Packaging Plastic bottle
Linear Formula C6H5C(CH3)2OOH
Quantity 2.5 kg
Specific Gravity 1.06
Solubility Information Solubility in water: miscible.
IUPAC Name 2-phenylpropane-2-peroxol
Formula Weight 152.19
Percent Purity 80%
Chemical Name or Material Cumyl hydroperoxide, 80%
Afficher plus Afficher moins
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Identifiant produit
  • Cumyl hydroperoxide
Mention d'avertissement
  • Danger
Catégorie de danger
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Organic peroxide Type E & F
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H242-Heating may cause a fire.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H304-May be fatal if swallowed and enters airways.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H373-May cause damage to organs.
Conseils de prudence
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Informations supplémentaires
  • MIXTURE LIST-Contain: Cumene hydroperoxide, Isopropylbenzene,Acetophenone

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.