missing translation for 'onlineSavingsMsg'
Learn More

Cumyl hydroperoxide, 80%, ACROS Organics™

Brand:  Acros Organics 349960010

Code : 29

Additional Details : CAS Number : 80-15-9 Weight : 1.05000kg Transport : UN number : 3109 Chem class : 5.2

Quantity 1kg
Packaging Plastic bottle

 View more versions of this product

Product Code. 10219373

  • £42.70 / 1kg


EU Stock 2
Add to basket

Description

Description

Oxidation reagent
Specifications

Chemical Identifiers

Cumyl hydroperoxide
98-86-2
C9H12O2
152.19
Cumene hydroperoxide, Trigonox(r) K
80%
80%
C6H5C(CH3)2OOH
MFCD00002129

Specifications

Cumyl hydroperoxide
1kg
98-86-2
Plastic bottle
80%
C6H5C(CH3)2OOH
MFCD00002129
Solubility in water: miscible.
83°C
80%
1.0600g/mL
80%
C9H12O2
152.19
Cumene hydroperoxide, Trigonox(r) K
− 30.0°C
Videos
SDS