missing translation for 'onlineSavingsMsg'
Learn More

Cumene, TRC

Product Code. 30004684
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004684 50 g 50g
30004618 10 g 10g
17943615 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004684 Supplier Toronto Research Chemicals Supplier No. TRCC83459050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-82-8
Molecular Formula C9 H12
Molecular Weight (g/mol) 120.19
Synonym Isopropylbenzene, Benzene, (1-methylethyl)-, Cumene (8CI), (1-Methylethyl)benzene, 2-Phenylpropane, Cumol, Isopropylbenzene, NSC 8776, i-Propylbenzene
IUPAC Name cumene
SMILES CC(C)c1ccccc1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C9H12/c1-8(2)9-6-4-3-5-7-9/h3-8H,1-2H3
Formula Weight 120.0939
Chemical Name or Material Isopropylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.