missing translation for 'onlineSavingsMsg'
Learn More

Cumene Hydroperoxide (80%, Technical grade), TRC

Product Code. 30366586
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30366586 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30366586 Supplier Toronto Research Chemicals Supplier No. TRCC8346005G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cumene Hydroperoxide (Technical grade)
CAS 80-15-9
Molecular Formula C9H12O2
Quantity 5 g
Synonym 1-Methyl-1-phenylethyl Hydroperoxide, alpha,alpha-Dimethylbenzyl Hydroperoxide, 1-Methyl-1-phenylethyl Hydroperoxide, 2-Hydroperoxy-2-phenylpropane, 2-Phenyl-2-propyl Hydroperoxide, 2-Phenylpropyl Hydroperoxide, 7-Cumyl Hydroperoxide, CHP, CHP 158, CHP 5, CHP 90, CHPO, CU 90, Cumen Hydroperoxide, Cumene Hydroperoxide, Cumenyl Hydroperoxide, Cumyl Hydrogen peroxide, Cumyl hydroperoxide, H 80, Hyperis, Hyperiz, Isopropylbenzene Hydroperoxide, Kayacumene H, Luperox CU 80, Luperox CU 90, PH 80, Percumyl H, Percumyl H 18, Percumyl H 80, Percumyl H 90, R 239A, Trigonox K 80, Trigonox K 90, Trigonox R 239A, Trigonox R 239R, alpha,alpha-Dimethylbenzyl Hydroperoxide, alpha-Cumene Hydroperoxide, alpha-Cumyl Hydroperoxide
InChI Formula InChI=1S/C9H12O2/c1-9(2,11-10)8-6-4-3-5-7-8/h3-7,10H,1-2H3
SMILES CC(C)(OO)c1ccccc1
IUPAC Name 2-hydroperoxypropan-2-ylbenzene
Molecular Weight (g/mol) 152.19
Formula Weight 152.0837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.