missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CultureCEPT™ Supplement (1000X)

Product Code. 17920995 Shop All Gibco Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
Unit Size:
0.1mL
0.5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17920995 0.1 mL 0.1mL
17938332 0.5 mL 0.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17920995 Supplier Gibco™ Supplier No. A56800
Cell Culture Essentials
Use promo code 27976
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gibco CultureCEPT Supplement reduces cellular stress and improves viability of stem cells, differentiated cells, and primary cells during a wide variety of handling and processing steps where cell damage and death can limit the success of a workflow.

Gibco CultureCEPT Supplement reduces cellular stress and improves viability of stem cells, differentiated cells, and primary cells during a wide variety of handling and processing steps where cell damage and death can limit the success of a cell culture workflow. It is a unique formulation that provides superior cell protection compared to traditional ROCK inhibitors, such as Y-27632, and other commercially available cell culture reagents.

The formulation is based on advancements from the Stem Cell Translation Laboratory at the NIH's National Center for Advancing Translational Sciences (NCATS), where a screen of over 15,000 compounds identified a combination of small molecules (referred to as CEPT) that work together to provide unmatched cytoprotection in passaging pluripotent stem cells, embryoid body creation, organoid formation, single-cell cloning, gene-editing, differentiation, and recovery of cryopreserved cells including PSCs, cardiomyocytes, neurons, astrocytes, and hepatocytes. CultureCEPT Supplement contains the CEPT cocktail molecules, chroman 1, emricasan, polyamines, and trans-ISRIB, to provide improved cell viability.

Achieve unparalleled cell survival in numerous applications

CultureCEPT Supplement will benefit scientists looking for:

  • Improved viability of PSCs, differentiated cells, or primary cells following recovery from cryopreservation
  • Improved viability during the passaging of stem cells
  • Improved viability during electroporation or transfection
  • Improved clonal efficiency during gene-editing and clonal selection with PSCs
  • Improved embryoid body and organoid formation

CultureCEPT Supplement harnesses the power of CEPT

  • Innovative small molecules comprising the CEPT cocktail have been extensively tested by the NIH's NCATS (Chen et al., 2021)
  • Growing number of publications highlights utility and exceptional performance of CEPT small molecules

Compatible with trusted Gibco media systems

  • Evaluated in StemScale PSC Suspension Medium, StemFlex Medium, Essential 8 Medium, Essential 8 Flex Medium, Neurobasal Medium, and Neurobasal Plus Medium
  • For over 60 years Gibco reagents have enabled successful cell culture

Ready-to-use

  • Fully formulated 1000X
  • Simply add directly to media
  • No weighing, no resuspending, no displacing media, no hassle

Citations and references

Chen Y, et al. A versatile polypharmacology platform promotes cytoprotection and viability of human pluripotent and differentiated cells. Nat Methods. 2021 May;18(5):528-541.

Tristan CA, et al. Efficient and safe single-cell cloning of human pluripotent stem cells using the CEPT cocktail. Nat Protoc. 2023 Jan;18(1):58-80.

Ryu S, et al. Stress-free cell aggregation by using the CEPT cocktail enhances embryoid body and organoid fitness. Biofabrication. 2023 Dec;16(1):10.1088/1758-5090/ad0d13.

Deng T, et al. Scalable generation of sensory neurons from human pluripotent stem cells. Stem Cell Reports. 2023 Apr 11;18(4):1030-1047.

Jovanovic VM, et al. A defined roadmap of radial glia and astrocyte differentiation from human pluripotent stem cells. Stem Cell Reports. 2023 Aug;18(8):1701-1720.

McIntire E, et al. Guided Differentiation of Pluripotent Stem Cells for Cardiac Cell Diversity. Preprint. bioRxiv. 2024 Feb;2023.07.21.550072.

Jin M, et al. Generation of iPSC-based human-mouse microglial brain chimeras to study senescence of human microglia. STAR Protoc. 2022 Dec;3(4):101847.

Liu L, et al. Modeling post-implantation stages of human development into early organogenesis with stem-cell-derived peri-gastruloids. Cell. 2023 Aug;186(18):3776-3792.e16.

Kang SY, et al. A Pillar and Perfusion Plate Platform for Robust Human Organoid Culture and Analysis. Adv Healthc Mater. Published online August 24, 2023.

Deng C, et al. Massively parallel characterization of psychiatric disorder-associated and cell-type-specific regulatory elements in the developing human cortex. Preprint. bioRxiv. 2023 Feb;2023.02.15.528663.

Tristan CA, et al. Robotic high-throughput biomanufacturing and functional differentiation of human pluripotent stem cells. Stem Cell Reports. 2021 Dec 14;16(12):3076-3092.

Ryu S, et al. Human Pluripotent Stem Cells for High-Throughput Drug Screening and Characterization of Small Molecules. Methods Mol Biol. 2022;2454:811-827.

Held A, Adler M, Marques C, et al. iPSC motor neurons, but not other derived cell types, capture gene expression changes in postmortem sporadic ALS motor neurons. Cell Rep. 2023 Sep;42(9):113046.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage –5 to –25°C. Protect from light.
Concentration 1000X
Culture Type Mammalian Cell Culture
Form Solution
Product Type Cell Culture Supplement
Sterility Sterile-filtered
Quantity 0.1 mL
Can CultureCEPT Supplement be used with routine passaging of PSCs long-term?

Published data shows that CEPT small molecules are suitable for long-term serial passaging of hPSCs. Normal pluripotency and karyotypes are observed after 40 passages (Chen Y, et al. A versatile polypharmacology platform promotes cytoprotection and viability of human pluripotent and differentiated cells. Nat Methods. 2021 May;18(5):528-541).

Can CultureCEPT Supplement effectively improve survival after cryopreservation if not included in the freezing medium?

Yes, CultureCEPT Supplement is fast acting and provides significant benefit to cells recovering from cryopreservation if introduced in the cell culture media during post-thaw recovery.

When using CultureCEPT Supplement, does seeding density need to be adjusted?

Seeding density should be examined when using CultureCEPT Supplement to ensure optimal conditions. In many cases, the appropriate seeding density is based on the number of viable cells in each plate or well. When achieving increased viability with CultureCEPT Supplement, you will need to account for this. In many scenarios, it may be possible to plate fewer cells.

What mechanisms are involved with CEPT small molecules present in CultureCEPT Supplement?

Chroman 1 is a ROCK2 inhibitor, Emricasan is a pan-caspase inhibitor, polyamines provide essential polycations, and trans ISRIB is an integrated stress response inhibitor.

Does CultureCEPT Supplement contain animal-derived components?

No, CultureCEPT Supplement only contains small molecules.

Once thawed, can CultureCEPT Supplement be stored in a refrigerator?

Long-term storage of CultureCEPT Supplement is recommended at -20 degrees C, where the supplement is stable for at least 12 months. After thawing, the supplement can be kept at 4 degrees C for 1 week.

Can I aliquot and re-freeze CultureCEPT Supplement?

Yes, CultureCEPT Supplement can be aliquoted and re-frozen. We recommend limiting to one re-freeze.

Will CultureCEPT Supplement work with different cell lines?

Yes, CultureCEPT Supplement has been evaluated on a variety of hPSC lines and different types of primary cells. Also, the small molecules of CEPT have been demonstrated to improve cell survival independent of the cell line (Chen Y, et al. A versatile polypharmacology platform promotes cytoprotection and viability of human pluripotent and differentiated cells. Nat Methods. 2021 May;18(5):528-541).

Does CultureCEPT Supplement require a specific medium?

No, CultureCEPT Supplement can work with a wide variety of media systems, such as Essential 8 Medium, Essential 8 Flex Medium, StemFlex Medium, StemScale PSC Suspension Medium, Neurobasal Plus Medium, and Neurobasal Medium.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.