Learn More
Eppendorf™ Culture Flask Adapter
Designed for use with Eppendorf Rotors A-4-81-MTP/Flex
Brand: Eppendorf™ 5825719000
Code : A0
Additional Details : Weight : 0.25600kg
Description
- Adapter
- For 1 cell culture flask
- For Rotors A-4-81-MTP/Flex
Specifications
Adapter | |
A-4-81-MTP/Flex and A-4-62-MTP | |
2 Pack |