missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cultrex Synthetic Hydrogel Kit (RUO), 1 mL

Product Code. 31794262
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
31794262 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31794262 Supplier R&D Systems™ Supplier No. CSHRUO01
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Extracellular Matrix for 3D Organoid Cultures - Fully Defined, Synthetic Component Derived Formulation.

Improved Culture Consistency: Controlled formulation reduces variables, Uncompromised Performance: Comparable performance to animal component derived matrices, Improved Imaging: Non-Fluorescent hydrogel is optimal for high- and super-resolution microscopy methods, Efficient Harvesting: Synthetic component derived formulation allows for facile organoid and cell extraction with protease digestion, User-Friendly Format: Kit format provided with validated protocol to support out-of-the-box user experience
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell Culture
Quantity 1 mL
Endotoxin Level ≤ 4 EU/mL by Limulus Amoebocyte Lysate (LAL) assay
Content And Storage Store the unopened product at -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.