missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Cultrex Stem Cell Qualified RGF Basement Membrane Extract
Click to view available options
Quantity:
1 mL
5 mL
Unit Size:
1mL
5mL
Description
Promotes the attachment of human iPSCs. Effectively maintains human iPSCs in a pluripotent state in a feeder-free culture.
Specifications
Specifications
| Concentration | 12 to 18mg/mL |
| For Use With (Application) | Bioactivity |
| Quantity | 1 mL |
| Endotoxin Level | ≤ 8EU/mL |
| Source | Murine Engelbreth-Holm-Swarm (EHS) tumor. |
| Content And Storage | Product is stable for a minimum of 3 months from date of shipment when stored at -20° C in a manual defrost freezer. For optimal stability, store at -80° C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction