missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cultrex ReadyBME, Stem Cell Qualified, Reduced Growth Factor

Product Code. 18401750
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18401750 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18401750 Supplier R&D Systems™ Supplier No. 3434050RTU
  • ONLINE EXCLUSIVE PRICE
    £99.32
    £88.20 / 50mL
    Save £11.12
    11% Off
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pre-diluted Cultrex Stem Cell-Qualified RGF Basement Membrane Extract

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Product Line R&D Systems
Quantity 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.