missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Reg1A Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems IC4937U-100UG
This item is not returnable.
View return policy
Description
Reg1A Monoclonal specifically detects Reg1A in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Reg1A | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P05451 | |
| Rat | |
| 100 μg | |
| Primary | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 431202 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| REG1A | |
| E. coli-derived recombinant human Reg1A Gln23-Asn166 Accession # P05451 | |
| RUO | |
| Detects human Reg1A in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) Reg1B, rhReg4, recombinant rat (rr) Reg2, or rrReg3. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction