missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human ICAM-1/CD54 Non Allele-specific Quantikine ELISA Kit

Product Code. 30001351
Click to view available options
Quantity:
1 Kit
6 Pack
Unit Size:
Pack of 96
This item is not returnable. View return policy

Product Code. 30001351

Brand: R&D Systems™ DCIM00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human ICAM-1/CD54 Non Allele-specific Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human ICAM-1/CD54 Non Allele-specific Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human ICAM-1/CD54 in cell culture supernates (100 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), urine (50 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.63 - 40ng/mL
Assay Sensitivity 0.053ng/mL
Assay Solid Phase Sandwich ELISA
Conjugate HRP
Cross Reactivity < 0.5% cross-reactivity observed with available related molecules. < 50% cross-species reactivity observed with species tested.
Immunoassay Kit Format 96-well Strip Plate
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine
Specificity Natural and Recombinant Human ICAM-1
Target Species Human
For Use With (Application) ELISA
Incubation Time 2.5 hr.
Quantity 1 Kit
Sample Volume Cell Culture Supernates (100uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Urine (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM1, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.