missing translation for 'onlineSavingsMsg'
Få mere at vide

Thermo Scientific™ CultiMaxx™ Shelving System for G-Rex 500M-CS Bioreactors

Artikelnummer. 17988084 Shop alle Thermo Scientific produkter
Skift visning
Klik for at se tilgængelige muligheder
No. of Shelves:
7
Material:
Stainless Steel
Pakningsstørrelse:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. No. of Shelves Material Pakningsstørrelse
17988084 7 Stainless Steel 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17988084 Leverandør Thermo Scientific™ Leverandørnr. 332395G08
  • ONLINE EXCLUSIVE PRICE
    £7595.20
    £5388.51 / 1 sets
    Save £2206.69
    29% Off
 Restricted Item
Estimated Shipment: 16-02-2027
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

CultiMaxx™ Shelving Systems to support G-Rex 500M-CS Bioreactors

Maximize the capacity, increase throughput and optimize cell growth in your lab using the CultiMaxx shelving system in a range of large capacity CO2 Incubators.

Maximize incubator space  

  • These shelving systems are designed to support scale-out of the G-Rex 500M-CS* in cell therapy production.
  • With an in-depth review of the shelving design, this new system provides up to a 150% increase in capacity for the G-Rex, or another way to look at it: 2 1/2 incubators in 1.

 Easy installation

  • Shelving systems are retrofittable and can be self-installed.
*G-Rex is a registered trademark of Wilson Wolf Corporation. G-Rex bioreactors must be ordered from Wilson Wolf Corporation.
TRUSTED_SUSTAINABILITY

Tekniske data

Description Shelving system to support G-Rex™ 500M for Reach-In 821L CO2 Incubator
For Use With (Equipment) Reach-In 821L CO2 Incubator
Material Stainless Steel
No. of Shelves 7
Type Shelf
Includes 7 levels of shelving
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.