Learn More
Thermo Scientific™ Culti-Loops™ Streptococcus thermophilus ATCC™ 19258™
Product Code. 13206179
Change view
Click to view available options
Quantity:
5 Loops/Pack
Unit Size:
Pack of 5
Product Code. Quantity unitSize
13206179 5 Loops/Pack Pack of 5
1 options
If you would like to order, a signed ATCC agreement is required.
Product Code. 13206179 Supplier Thermo Scientific™ Supplier No. R4608238
  • ONLINE EXCLUSIVE PRICE
    £197.00
    £192.75 / Pack of 5
    Save £4.25
    2% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
If you would like to order, a signed ATCC agreement is required.
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 19258TM*
Description Streptococcus thermophilus ATCC™ 19258™
Product Type Culti-Loops Quality Control Organism
Target Organism Class Cocci
Test Type Qualitative QC
Quantity 5 Loops/Pack
Strain Designations WDCM00023, NCDO 573 [NCIMB 8510, B of R, CNCTC 28/89, DSM 20617, CCUG 21957, CIP 102303, LMG 6896]
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.