Learn More
Thermo Scientific™ Culti-Loops™ Staphylococcus aureus NCTC 12493™
Description
Confidently carry out testing of media, reagents, and identification & antimicrobial susceptibility kits and evaluate bacteriological procedures with ready-to-use Thermo Scientific™ Culti-Loops™ QC organisms.
Staphylococcus aureus NCTC 12493™ is methicillin resistant (MRSA) conferred by mec-A gene. EUCAST recommends this strain to be tested as part of extended QC for cefoxitin disc testing.
Technique:
The loop can be streaked directly onto appropriate media.
Alternatively, it may be dissolved in a liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000.
Includes:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
Specifications
| Description | Staphylococcus aureus NCTC 12493™ |
| Format | Culti-Loops™ |
| Target Organism Class | Gram-positive Cocci |
| Test Type | Qualitative QC |
| NCTC® | NCTC 12493™ |
| Quantity | 5 Loops/Pk. |
| Strain Designations | MQCL 113, WDCM 00212 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.