Learn More
Thermo Scientific™ Culti-Loops™ Sporothrix schenckii ATCC™ 10212™
Product Code. 12985853 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Culti-Loops Quality Control Organism
Unit Size:
Pack of 5
Product Code. Product Type unitSize
12985853 Culti-Loops Quality Control Organism Pack of 5
1 options
If you would like to order, a signed ATCC agreement is required.
Product Code. 12985853 Supplier Thermo Scientific™ Supplier No. R4608205
If you would like to order, a signed ATCC agreement is required.
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 10212TM*
Description Sporothrix schenckii ATCC™ 10212™
Product Type Culti-Loops Quality Control Organism
Target Organism Class Fungi
Test Type Qualitative QC
Packaging Type 5 Loops/Package
Strain Designations NIH 7019
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.