Learn More
Thermo Scientific™ Culti-Loops™ Sensititre™ H. influenzae/S.pneumoniae MIC QC Set
Description
Thermo Scientific™ Culti-Loops™ Sensititre H. influenzae/S.pneumoniae MIC Set contains quality control organisms supplied in Culti-Loops format.
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Includes:
Haemophilus influenzae ATCC™ 49247™ †
Haemophilus influenzae ATCC™ 49766™†
Streptococcus pneumoniae ATCC™ 49619™ †
Ordering Information:
Individually sealed in a foil pouch and packaged five loops per package.
Specifications
Specifications
| ATCC® | 49247TM* |
| Description | Sensititre™ H. influenzae/S.pneumoniae MIC QC Set/3 |
| Format | Culti-Loops |
| Product Type | QC set |
| Target Organism Class | Organism Sets |
| Test Type | Qualitative QC |
| Quantity | 3 x 5 Loops |
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.