Learn More
Thermo Scientific™ Culti-Loops™ Sensititre™ Automated Gram Negative QC Set
Description
Thermo Scientific™ Culti-Loops™ Sensititre Automated Gram Negative Set contains quality control organisms supplied in Culti-Loops format.
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Includes:
Edwardsiella tarda ATCC™ 15947 ™†
Klebsiella oxytoca ATCC™ 8724™ †
Morganella morganii subsp. morganii ATCC™ 25830™ †
Proteus vulgaris ATCC™ 6896™ †
Pseudomonas aeruginosa ATCC™ 10145 ™†
Shigella sonnei ATCC™ 25931™ †
Specifications
Specifications
| ATCC® | 15947TM* |
| Description | Sensititre™ Automated Gram Negative QC Set/6 |
| Format | Culti-Loops |
| Product Type | QC set |
| Target Organism Class | Organism Sets |
| Test Type | Qualitative QC |
| Packaging Type | 6 Loops/Pack |
| Quantity | 6 x 1 Loops |
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.