missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Culti-Loops™ Rhizopus stolonifer ATCC™ 14037™

Product Code. 12925833
Click to view available options
Product Type:
Culti-Loops Quality Control Organism
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 12925833

Brand: Thermo Scientific™ R4607212

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.

Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.

The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.

Ordering Information:

Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.

TRUSTED_SUSTAINABILITY

Specifications

ATCC® 14037TM*
Description Rhizopus stolonifer ATCC™ 14037™
Format Culti-Loops
Product Type Culti-Loops Quality Control Organism
Target Organism Class Fungi
Test Type Qualitative QC
Quantity 5 Loops/Pack
Strain Designations UNB-1 [IMI 90609, VKM F-402]

*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.