missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Culti-Loops™ Porphyromonas gingivalis ATCC™ 33277™

Product Code. 12925893
Change view
Click to view available options
Quantity:
5 Loops/Pack
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12925893 5 Loops/Pack Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
If you would like to order, a signed ATCC agreement is required.
Product Code. 12925893 Supplier Thermo Scientific™ Supplier No. R4609008
  • ONLINE EXCLUSIVE PRICE
    £258.16
    £235.73 / Pack of 5
    Save £22.43
    8% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy
If you would like to order, a signed ATCC agreement is required.

Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.

Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.

The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.

Ordering Information:

Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.

TRUSTED_SUSTAINABILITY

Specifications

ATCC® 33277TM*
Description Porphyromonas gingivalis ATCC™ 33277™
Format Culti-Loops
Product Type Culti-Loop
Target Organism Class Anaerobes
Quantity 5 Loops/Pack

*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.