Learn More
Thermo Scientific™ Culti-Loops™ Klebsiella aerogenes ATCC™ 13048™
Product Code. 12945803
Change view
Click to view available options
Quantity:
5 Loops/Pack
Unit Size:
Pack of 5
Product Code. Quantity unitSize
12945803 5 Loops/Pack Pack of 5
1 options
If you would like to order, a signed ATCC agreement is required.
Product Code. 12945803 Supplier Thermo Scientific™ Supplier No. R4607080
If you would like to order, a signed ATCC agreement is required.
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 13048TM*
Description Klebsiella aerogenes ATCC™ 13048™
Product Type Culti-Loops Quality Control Organism
Target Organism Class Gram-negative Rods
Test Type Qualitative QC
Quantity 5 Loops/Pack
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.