Learn More
Thermo Scientific™ Culti-Loops™ Corynebacterium urealyticum ATCC™ 43044™
Product Code. 12945953
Change view
Click to view available options
Quantity:
5 Loops/Pk.
Unit Size:
Pack of 5
Product Code. Quantity Unit Size
12945953 5 Loops/Pk. Pack of 5
1 options
If you would like to order, a signed ATCC agreement is required.
Product Code. 12945953 Supplier Thermo Scientific™ Supplier No. R4609414
Reliable Agri Testing Solutions
Use promo code 28024
View all promotions
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
If you would like to order, a signed ATCC agreement is required.
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 43044TM*
Certifications/Compliance Culti-Loops are CE Marked.
Description Corynebacterium urealyticum
Product Type Culti-Loops Quality Control Organism
Target Organism Class Gram-positive Rods
Test Type Qualitative QC
Packaging Type 5 Loops/Pack
Quantity 5 Loops/Pk.
Strain Designations WDCM00211, 3 [Garcia strain]
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.