Learn More
Thermo Scientific™ Culti-Loops™ Corynebacterium diphtheriae ATCC™ 13812™
Product Code. 12905923
Change view
Click to view available options
Quantity:
5 Loops/Pk.
Unit Size:
Pack of 5
Product Code. Quantity unitSize
12905923 5 Loops/Pk. Pack of 5
1 options
If you would like to order, a signed ATCC agreement is required.
Product Code. 12905923 Supplier Thermo Scientific™ Supplier No. R4609234
If you would like to order, a signed ATCC agreement is required.
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 13812TM*
Certifications/Compliance Culti-Loops are CE Marked.
Description Corynebacterium diphtheriae
Product Type Single Organism
Target Organism Class Gram-positive Rods
Test Type Qualitative QC
Packaging Type 5 Loops/Pack
Quantity 5 Loops/Pk.
Strain Designations WDCM00054
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.