Learn More
Thermo Scientific™ Culti-Loops™ Bacillus megaterium ATCC™ 14581™
Product Code. 12995943
Change view
Click to view available options
Quantity:
5 Loops
Unit Size:
Pack of 5
Product Code. Quantity Unit Size
12995943 5 Loops Pack of 5
1 options
If you would like to order, a signed ATCC agreement is required.
Product Code. 12995943 Supplier Thermo Scientific™ Supplier No. R4609395
  • ONLINE EXCLUSIVE PRICE
    £210.00
    £185.26 / Pack of 5
    Save £24.74
    11% Off
 Restricted Item
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
If you would like to order, a signed ATCC agreement is required.
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 14581TM*
Description Bacillus megaterium
Product Type Culti-Loops Quality Control Organism
Target Organism Class Gram-positive Rods
Test Type Qualitative QC
Packaging Type 5 Loops/Pack
Quantity 5 Loops
Strain Designations WDCM00004, [BCRC 10608, CCM 2007, CCUG 1817, CIP 66.20, DSM 32, HAMBI 2018, IAM 13418, JCM 2506, KCTC 3007, LMG 7127, NBRC 15308, NCCB 75016, NCIMB 9376, NCTC 10342, NRIC 1710, NRRL B-14308, VKM B-512]
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.