Learn More
Thermo Scientific™ Culti-Loops™ Achromobacter xylosoxidans subsp. xylosoxidans ATCC™ 27061™
Description
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
Specifications
| ATCC® | 27061TM* |
| Certifications/Compliance | Culti-Loops are CE Marked. |
| Description | Achromobacter xylosoxidans subsp. Xylosoxidans |
| Product Type | Culti-Loops Quality Control Organism |
| Target Organism Class | Gram-negative Rods |
| Test Type | Qualitative QC |
| Packaging Type | 5 Loops/Pack |
| Quantity | 5 Loops/Pk. |
| Strain Designations | KM 543 [CCUG 12689, CIP 71.32, DSM 10346, DSM 2402, EY 543, GIFL 543, IFO 15126, JCM 9659, LMG 1863, NCTC 10807, NRRL B-4082, R. Hugh 2838] |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.