missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cullin-7 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-602B
This item is not returnable.
View return policy
Description
Cullin-7 Polyclonal antibody specifically detects Cullin-7 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Cullin-7 | |
| Polyclonal | |
| Western Blot | |
| CUL-7, cullin 7, cullin-7, KIAA0076dJ20C7.5 | |
| A synthetic peptide, mapping to a region between residue 1650 and the C-terminus (residue 1698) of human Cullin homolog 7 using the numbering given in SwissProt entry Q14999 (GeneID 9820). | |
| 0.1 mL | |
| Angiogenesis, DNA Repair | |
| 9820 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction