missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cullin 5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92652-0.02ml
This item is not returnable.
View return policy
Description
Cullin 5 Polyclonal antibody specifically detects Cullin 5 in Rat samples. It is validated for Western Blot
Specifications
| Cullin 5 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:5000 | |
| CUL-5, cullin 5, VACM-1cullin-5, VACM1Cullin-5 (vasopressin-activated calcium-mobilizing receptor-1), Vasopressin-activated calcium-mobilizing receptor 1, Vasopressin-activated calcium-mobilizing receptor-1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human CUL5 (NP_003469.2). MATSNLLKNKGSLQFEDKWDFMRPIVLKLLRQESVTKQQWFDLFSDVHAVCLWDDKGPAKIHQALKEDILEFIKQAQARVLSHQDDTALLKAYIVEWRKFFTQCDILPKPFCQLEITLMGKQGSNKKSNVEDSIVRKLMLDTWNESIFSNIKNRLQDSAMKLVHAERLGEAFDSQLVIGVRESYVNLCSNPEDKLQIYRDNFEKAYLDSTERFYRTQAPSYLQQNGVQNYMKYADAKLKEEEKRALRYLETRRECNSVEALMECCVNALVTSFKETILAECQGMIKRNETEKLHLMFSLM | |
| 0.02 mL | |
| Cancer | |
| 8065 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction