missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cullin 4a Antibody (5A7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42951
This item is not returnable.
View return policy
Description
Cullin 4a Monoclonal antibody specifically detects Cullin 4a in Human,Monkey,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Cullin 4a | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Monkey, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 5A7 | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry 1:20 - 1:200, Immunocytochemistry/ Immunofluorescence 1:20 - 1:200, Immunoprecipitation 1:500 - 1:1000 | |
| CUL-4A, cullin 4A, cullin-4A | |
| cullin 4A - (Uniprot# Q13619) | |
| 100 μg | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 8451 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction