Learn More
Cullin 2 Recombinant Rabbit Monoclonal Antibody (50H17L12), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen 700179
Code : NEW
Additional Details : Weight : 1.00000kg
Description
This antibody is predicted to react with bovine, canine, chimpanzee, equine, orangutan, primate, Rhesus monkey and Xenopus based on sequence homology. Intact IgG appears on a non-reducing gel as ∽150 kDa band and upon reduction generating a ∽25 kDa light chain band and a ∽50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Core component of multiple cullin-RING-based ECS (ElonginB/C-CUL2/5-SOCS-box protein) E3 ubiquitin-protein ligase complexes, which mediate the ubiquitination of target proteins. ECS complexes and ARIH1 collaborate in tandem to mediate ubiquitination of target proteins (PubMed:27565346). May serve as a rigid scaffold in the complex and may contribute to catalysis through positioning of the substrate and the ubiquitin-conjugating enzyme. The E3 ubiquitin-protein ligase activity of the complex is dependent on the neddylation of the cullin subunit and is inhibited by the association of the deneddylated cullin subunit with TIP120A/CAND1. The functional specificity of the ECS complex depends on the substrate recognition component. ECS(VHL) mediates the ubiquitination of hypoxia-inducible factor (HIF). [UniProt]Specifications
Cullin 2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q13617, Q9D4H8 | |
Cul2 | |
A peptide corresponding to amino acids 733-745 of Q13617. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
50H17L12 | |
Unconjugated | |
Cul2 | |
1300003D18Rik; 4932411N15Rik; AI327301; CUL2; CUL-2; cullin 2; cullin-2; MGC131970; mKIAA4106; OTTHUMP00000019437; testis secretory sperm-binding protein Li 238E | |
Rabbit | |
Protein A | |
RUO | |
361258, 71745, 8453 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |