missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Cullin 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29236
This item is not returnable.
View return policy
Description
Cullin 2 Polyclonal antibody specifically detects Cullin 2 in Human,Mouse samples. It is validated for Immunohistochemistry
Specifications
| Cullin 2 | |
| Polyclonal | |
| Tris-buffered Saline 0.1% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry | |
| Unconjugated | |
| CUL-2, cullin 2, cullin-2, MGC131970 | |
| Between 150 and 200 | |
| 10 μg | |
| Apoptosis, Cancer, Cell Cycle and Replication, DNA Repair, DNA replication Transcription Translation and Splicing | |
| 8453 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction