missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cullin 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-16092-100UL
This item is not returnable.
View return policy
Description
Cullin 1 Polyclonal antibody specifically detects Cullin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
| Cullin 1 | |
| Polyclonal | |
| PBS, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| CUL-1, cullin 1, cullin-1, MGC149834, MGC149835 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 435-593 of human Cullin 1 (NP_003583.2). | |
| 100 μg | |
| Apoptosis, Cell Cycle and Replication | |
| 8454 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction