missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUL7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13738-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a component of an E3 ubiquitin-protein ligase complex. The encoded protein interacts with TP53, CUL9, and FBXW8 proteins. Defects in this gene are a cause of 3M syndrome type 1 (3M1). Two transcript variants encoding different isoforms have been found for this gene.Specifications
| CUL7 | |
| Polyclonal | |
| Unconjugated | |
| CUL7 | |
| CUL 7, CUL7, cullin 7, KIAA0076 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 66515, 9820 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14999, Q8VE73 | |
| Cul7 | |
| CUL7 Fusion Protein Ag4675 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction