missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUL4B Antibody (OTI1C4), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71373AF350
This item is not returnable.
View return policy
Description
CUL4B Monoclonal specifically detects CUL4B in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| CUL4B | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CUL4B | |
| Recombinant protein fragment corresponding to amino acids 1-300 of human CUL4B (NP_003579) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI1C4 | |
| Western Blot, Immunohistochemistry | |
| CUL-4B, cullin 4B, cullin-4B, DKFZp686F1470, KIAA0695, MRXHF2, MRXS15, MRXSC, SFM2 | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication | |
| 8450.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction