missing translation for 'onlineSavingsMsg'
Learn More

CUL4A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14851-1-AP-150UL

 View more versions of this product

Product Code. 16873714

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is specifically against CUL4A,but not CUL4B. CUL4A has 2 isoforms with MW 88 kDa (759 aa) and 77 kDa (659 aa), and 14851-1-AP recognizes both.

CUL4A (Cullin 4A) is the ubiquitin ligase component of a multimeric complex involved in the degradation of DNA damage-response proteins.
Specifications

Specifications

CUL4A
Polyclonal
Unconjugated
CUL4A
CUL 4A, CUL4A, cullin 4A
Rabbit
Antigen Affinity Chromatography
RUO
361181, 8451, 99375
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q13619, Q3TCH7
Cul4a
Peptide
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.