missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUL4A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14851-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specifically against CUL4A,but not CUL4B. CUL4A has 2 isoforms with MW 88 kDa (759 aa) and 77 kDa (659 aa), and 14851-1-AP recognizes both.
CUL4A (Cullin 4A) is the ubiquitin ligase component of a multimeric complex involved in the degradation of DNA damage-response proteins.Specifications
| CUL4A | |
| Polyclonal | |
| Unconjugated | |
| CUL4A | |
| CUL 4A, CUL4A, cullin 4A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 361181, 8451, 99375 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13619, Q3TCH7 | |
| Cul4a | |
| Peptide | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction