missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUL2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10981-2-AP-20UL
This item is not returnable.
View return policy
Description
Stem cells offer a therapeutic promise for many diseases. In the case of diabetes, stem cells offer the promise that beta cells may be generated ex vivo for the possible transplantation and cure of Type I diabetes (Couri CE, et al.). For this to happen, stem cells will need to be isolated and expanded, they will need to be differentiated toward the beta cell lineage, and the differentiated cell progeny used for transplantation may need to be isolated from complex cell mixtures following differentiation. Appropriate markers targeting cell surface molecules on developing and mature pancreatic cells will be required for such research.Specifications
| CUL2 | |
| Polyclonal | |
| Unconjugated | |
| CUL2 | |
| CUL 2, CUL2, cullin 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8453 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q13617 | |
| CUL2 | |
| CUL2 Fusion Protein Ag1434 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction