Learn More
CUGBP2 Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12921-1-AP-20UL
 View more versions of this product
Product Code. 16890874
  • £135.00 / 20µL
Description
CUG-BP1 and CUG-BP2 are proteins that bind specifically to (CUG)8 oligonucleotides in vitro. While CUG-BP1 has the major binding activity in normal cells, nuclear CUG-BP2 binding activity increases in DM cells. Both CUG-BP1 and CUG-BP2 are isoforms of a novel heterogeneous nuclear ribonucleoprotein (hnRNP), hNab50. CUG-BP1, an RNA CUG triplet repeat binding protein, regulates splicing and translation of various RNAs. Expansion of RNA CUG repeats in the DMPK in DM is associated with alterations in binding activity of CUG-BP1 as well as alterations in the translation of the C/EBPb transcription factor. CUG-BP1 is an important regulator of initiation from different AUG codons of C/EBPb mRNA.
Specifications
Specifications
CUGBP2
Polyclonal
Unconjugated
CELF2
Bruno like protein 3, BRUNOL3, CELF 2, CELF2, CUG BP2, CUGBP2, ETR 3, ETR3, hNAPOR, NAPOR, RNA binding protein BRUNOL 3
Rabbit
Antigen Affinity Chromatography
RUO
10659, 14007, 29428, 373117
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation
0.51 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O95319, Q6P0B1, Q792H5, Q9Z0H4
celf2
CUGBP2 Fusion Protein Ag3594
20 μL
Primary
Human, Rat, Zebrafish, Mouse