missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUGBP2 Antibody (PCRP-CELF2-1E4), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14271PCP
This item is not returnable.
View return policy
Description
CUGBP2 Monoclonal antibody specifically detects CUGBP2 in Human samples. It is validated for Flow Cytometry
Specifications
| CUGBP2 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| Flow Cytometry | |
| PCRP-CELF2-1E4 | |
| Flow Cytometry | |
| BRUNOL3ELAV-type RNA-binding protein 3, Bruno-like protein 3, CELF-2, CUG triplet repeat RNA-binding protein 2, CUG triplet repeat, RNA binding protein 2, CUG triplet repeat, RNA-binding protein 2, CUG-BP- and ETR-3-like factor 2, CUGBP Elav-like family member 2, CUGBP, Elav-like family member 2, CUG-BP2, CUGBP2hNAPOR, Etr-3, ETR3RNA-binding protein BRUNOL-3, NAPOR, NAPOR-2, Neuroblastoma apoptosis-related RNA-binding protein | |
| Recombinant full-length human CUGBP2 protein (Uniprot: O95319) | |
| 0.1 mL | |
| Lipid and Metabolism, Transcription Factors and Regulators | |
| 10659 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction