missing translation for 'onlineSavingsMsg'
Learn More

CUGBP2 Antibody (PCRP-CELF2-1E4), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18341835 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18341835 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18341835 Supplier Novus Biologicals Supplier No. NBP314271JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CUGBP2 Monoclonal antibody specifically detects CUGBP2 in Human samples. It is validated for Flow Cytometry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CUGBP2
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone PCRP-CELF2-1E4
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias BRUNOL3ELAV-type RNA-binding protein 3, Bruno-like protein 3, CELF-2, CUG triplet repeat RNA-binding protein 2, CUG triplet repeat, RNA binding protein 2, CUG triplet repeat, RNA-binding protein 2, CUG-BP- and ETR-3-like factor 2, CUGBP Elav-like family member 2, CUGBP, Elav-like family member 2, CUG-BP2, CUGBP2hNAPOR, Etr-3, ETR3RNA-binding protein BRUNOL-3, NAPOR, NAPOR-2, Neuroblastoma apoptosis-related RNA-binding protein
Host Species Mouse
Immunogen Recombinant full-length human CUGBP2 protein (Uniprot: O95319)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 10659
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.