missing translation for 'onlineSavingsMsg'
Learn More

CUGBP2 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18430314 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430314 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430314 Supplier Novus Biologicals Supplier No. NB600204MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CUGBP2 Polyclonal antibody specifically detects CUGBP2 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CUGBP2
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias BRUNOL3ELAV-type RNA-binding protein 3, Bruno-like protein 3, CELF-2, CUG triplet repeat RNA-binding protein 2, CUG triplet repeat, RNA binding protein 2, CUG triplet repeat, RNA-binding protein 2, CUG-BP- and ETR-3-like factor 2, CUGBP Elav-like family member 2, CUGBP, Elav-like family member 2, CUG-BP2, CUGBP2hNAPOR, Etr-3, ETR3RNA-binding protein BRUNOL-3, NAPOR, NAPOR-2, Neuroblastoma apoptosis-related RNA-binding protein
Host Species Rabbit
Immunogen C-terminus of human and mouse ETR-3
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 10659
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.