missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CUGBP1 Recombinant Rabbit Monoclonal Antibody (24GB3725)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557007
This item is not returnable.
View return policy
Description
CUGBP1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Myotonic dystrophy (MD) is an autosomal dominant neuromuscular disease that is associated with a (CTG)n repeat expansion in the 3'-untranslated region of the myotonin protein kinase (Mt-PK) gene. A (CUG) n oligonucleotides triplet repeat pre-mRNA/mRNA binding protein may play an important role in DM pathogenesis. HeLa cell protein, CUG-BP1, has been purified based upon its ability to bind specifically to (CUG) 8 oligonucleotides in vitro. CUG-BP1 is the major (CUG) 8 binding activity in normal cells. CUG-BP1 has been identified as isoforms of a novel heterogeneous nuclear ribonucleoprotein (hnRNP), hNab50. The CUG-BP/hNab50 protein is localized predominantly in the nucleus and is associated with polyadenylated RNAs in vivo. In vitro RNA-binding/photocrosslinking studies demonstrate that CUG-BP/hNab50 binds to RNAs containing the Mt-PK 3-UTR. The (CUG) n repeat region in Mt-PK mRNA is a binding site for CUG-BP/hNab50 in vivo, and triplet repeat expansion leads to sequestration of this hnRNP on mutant Mt-PK transcripts.
Specifications
| CUGBP1 | |
| Recombinant Monoclonal | |
| 0.40 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P28659, Q4QQT3, Q92879 | |
| CELF1 | |
| A synthesized peptide derived from human CELF1 (aa 1-45). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB3725 | |
| Unconjugated | |
| CELF1 | |
| 1600010O03Rik; 50 kDa nuclear polyadenylated RNA-binding protein; AA407467; Brain protein F41; Brunol2; bruno-like 2; bruno-like protein 2; Celf1; CELF-1; CUG RNA-binding protein; CUG triplet repeat RNA-binding protein 1; CUG triplet repeat, RNA binding protein 1; CUG triplet repeat, RNA-binding protein 1; Cugbp; CUG-BP; CUG-BP- and ETR-3-like factor 1; CUGBP Ela; CUGBP Elav-like family member 1; CUGBP, Elav-like family member 1; CUGBP, Elav-like family member 1-like; CUGBP1; CUG-BP1; D2Wsu101e; Deadenylation factor CUG-BP; deadenylation factor EDEN-BP; EDEN-BP; EDEN-BP homolog; embryo deadenylation element binding protein; embryo deadenylation element-binding protein homolog; HNAB50; NAB50; NAPOR; nuclear polyadenylated RNA-binding protein, 50-kD; RNA-binding protein BRUNOL-2; RP23-147D3.7 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10658, 13046, 362160 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction