missing translation for 'onlineSavingsMsg'
Learn More

CUGBP1/CELF1 Antibody (SR1914), Novus Biologicals™

Rabbit Monoclonal Antibody

£257.00 - £341.00

Specifications

Antigen CUGBP1/CELF1
Clone SR1914
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18653385
View Documents
Novus Biologicals
NBP3-22304-100ul
100 μg
£341.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18660784
View Documents
Novus Biologicals
NBP3-22304-50ul
50 μg
£257.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CUGBP1/CELF1 Monoclonal antibody specifically detects CUGBP1/CELF1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Spécification

Spécification

CUGBP1/CELF1
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50
Monoclonal
Purified
RUO
Human, Mouse, Rat
BRUNOL250 kDa nuclear polyadenylated RNA-binding protein, bruno-like 2, Bruno-like protein 2, CUG RNA-binding protein, CUG triplet repeat, RNA binding protein 1, CUG triplet repeat, RNA-binding protein 1, CUG-BP, CUG-BP- and ETR-3-like factor 1, CUGBP Elav-like family member 1, CUGBP, Elav-like family member 1, CUGBP1CUG triplet repeat RNA-binding protein 1, CUGBPCELF-1, Deadenylation factor CUG-BP, EDEN-BP, EDEN-BP homolog, embryo deadenylation element binding protein, Embryo deadenylation element-binding protein homolog, hNab50, NAB50CUG-BP1, NAPOR, nuclear polyadenylated RNA-binding protein, 50-kD, RNA-binding protein BRUNOL-2
A synthesized peptide derived from human CUGBP1/CELF1 (Uniprot #: Q92879)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
SR1914
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
Rabbit
Cancer, Chromatin Research, DNA replication Transcription Translation and Splicing, Neurodegeneration, Phospho Specific
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
10658
IgG
Affinity purified
Vidéos
FDS
Documents

Documents

Product Certifications
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis