missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CUG-BP2 Monoclonal Antibody (9B1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA542411
This item is not returnable.
View return policy
Description
Positive Control: PC-3M cell lysates, rat skeletal muscle tissue, mouse colon tissue, Daudi. Subcellular Location: Cytoplasm, Nucleus.
CUG-BP1 and CUG-BP2 are proteins that bind specifically to (CUG)8 oligonucleotides in vitro. While CUG-BP1 has the major binding activity in normal cells, nuclear CUG-BP2 binding activity increases in DM cells. Both CUG-BP1 and CUG-BP2 are isoforms of a novel heterogeneous nuclear ribonucleoprotein (hnRNP), hNab50. CUG-BP1, an RNA CUG triplet repeat binding protein, regulates splicing and translation of various RNAs. Expansion of RNA CUG repeats in the DMPK in DM is associated with alterations in binding activity of CUG-BP1 as well as alterations in the translation of the C/EBPb transcription factor. CUG-BP1 is an important regulator of initiation from different AUG codons of C/EBPb mRNA.
Specifications
| CUG-BP2 | |
| Monoclonal | |
| 2 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| O95319, Q792H5, Q9Z0H4 | |
| CELF2 | |
| Synthetic peptide within Human CELF2 aa 460-508. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 9B1 | |
| Unconjugated | |
| CELF2 | |
| B230218O03; B230345P09Rik; BRUNOL3; Bruno-like protein 3; C88023; cb906; CELF 2; Celf2; CELF-2; CUG triplet repeat RNA-binding protein 2; CUG triplet repeat, RNA binding protein 2; CUG-BP- and ETR-3-like factor 2; Cugbp- and Etr3-like factor 2; CUGBP Elav-like family member 2; CUGBP, Elav-like family member 2; Cugbp2; CUG-BP2; cugbp2 protein; D230046B21Rik; elav-type ribonucleoprotein-3; elav-type RNA-binding protein 3; ETR3; ETR-3; fj87b07; hNAPOR; mETR-3; mNapor; NAPOR; napor protein isoform; napor protein long isoform; napor protein short isoform; Napor-2; neur; neuroblastoma apoptosis-related RNA-binding protein; RNA-binding protein; RNA-binding protein Brunol3; RNA-binding protein BRUNOL-3; rNapor; RP23-124P7.1; wu:fj87b07 | |
| Mouse | |
| Protein G | |
| RUO | |
| 10659, 14007, 29428 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction