missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUEDC2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92138-0.1ml
This item is not returnable.
View return policy
Description
CUEDC2 Polyclonal antibody specifically detects CUEDC2 in Rat samples. It is validated for Western Blot
Specifications
| CUEDC2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| bA18I14.5, C10orf66, chromosome 10 open reading frame 66, CUE domain containing 2, CUE domain-containing protein 2, MGC2491 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human CUEDC2 (NP_076945.2). MELERIVSAALLAFVQTHLPEADLSGLDEVIFSYVLGVLEDLGPSGPSEENFDMEAFTEMMEAYVPGFAHIPRGTIGDMMQKLSGQLSDARNKENLQPQSSGVQGQVPISPEPLQRPEMLKEETRSSAAAAADTQDEATGAEEELLPGVD | |
| 0.1 mL | |
| Signal Transduction | |
| 79004 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction